Learn More
Corning™ Straw Pipets, Polypropylene, Sterile
GREENER_CHOICE
Decrease cross contamination risks using disposable pipets
Brand:  Corning™ EPI24B-01
 View more versions of this product
Product Code. 11825380
  • 366.15 € / Pack of 6000
Estimated Shipment: 31-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Product traceability on each bag
  • Sterile 10-3
  • Packaged individually in tamper evident single zip bags
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Polypropylene
6000/Cs.
2.2 mL
Straw Pipet
Sterile
25/Bag
24 cm
Single-use