missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gosselin™ Straight Container
Corning™ Gosselin™ Straight Container, Specimen Vials, PP come in different sizes and quantities, and are available sterile and non-sterile.
Brand: Gosselin™ TP52C-001
120.93 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Clear and break-resistant polypropylene (PP)
- Available in different sizes and quantities
- Color-coded screw caps
Specifications
Polypropylene | |
125 mL | |
380/Cs. |
Sterile | |
Blue | |
380/Pk. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction