Learn More
Gosselin™ T-shaped Polystyrene Spreader
This product is sterilized using an irradiation method compliant with the requirements of ANSI/AAMI/ISO 11137 to achieve a Sterility Assurance Level (SAL) of 10-3.
Brand:  Gosselin ETAT-05
Product Code. 12652285
  • 786.22 € / Pack of 4500
Estimated Shipment: 13-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Polystyrene (PS)
  • Packed in resealable bag
  • Curved end or 'T' end
  • Sterile
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Single-use
148 mm
Sterile
Green
T-Shape
For Dairy, Environmental, Industrial, Microbiological Uses