missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gosselin™ White Polystyrene Stirrer
missing translation for 'orderingAttributeHoverText'
Length (Metric):
150 mm
missing translation for 'unitSize'
Pack of 1000
Description
- Polystyrene (PS)
- Packed in tamper-evident resealable bag
Specifications
Specifications
| Sterility | Sterile |
| Color | White |
| Certifications/Compliance | EC N°1935/2004, ISO 11137 |
| Product Type | Stirrer |
| For Use With (Application) | Used in Dairy, Environment, Industries, Microbiology |
| Material | Polystyrene |
| Length (Metric) | 150 mm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction