missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GPR3 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20847-1-AP-20UL
This item is not returnable.
View return policy
Description
The G-protein-coupled receptor (GPCR) superfamily is comprised of an estimated 600-1, 000 members and is the largest known class of molecular targets with proven therapeutic value. GPR3 is an orphan receptor with constitutive G signaling activity. GPR3 modulates a number of brain functions, including behavioral responses to stress, neurite outgrowth and amyloid-beta peptide generation suggesting that it may play a role in Alzheimer′s disease.Specifications
GPR3 | |
Polyclonal | |
Unconjugated | |
GPR3 | |
ACCA, ACCA orphan receptor, G protein coupled receptor 3, GPR3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
2827 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.27 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P46089 | |
GPR3 | |
GPR3 Fusion Protein Ag14257 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |