Learn More
GPX4 Mouse anti-Human, CoraLite 488, Clone: 3F5G5, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL488-67763-100UL
This item is not returnable.
View return policy
Description
GPX4 (Phospholipid hydroperoxide glutathione peroxidase, mitochondrial) protects cells against membrane lipid peroxidation and cell death. Required for normal sperm development and male fertility.It has two isforms about 20KDa and 22KDa, respectively. GPX4 is a monomer,but it has a tendency to form higher mass oligomers (PMID:17630701). It presents primarily in testis.
Specifications
GPX4 | |
Monoclonal | |
1000 μg/mL | |
Mouse | |
Primary | |
Human, Non-human Primate | |
IgG2b |
Immunofluorescence | |
3F5G5 | |
CoraLite 488 | |
100 μL | |
2879 | |
Liquid |