Learn More
Grace™ Jones Genesis™ C18 Reversed Phase HPLC Columns
A unique proprietary end-capping reagent provides freedom from residual silanols and enhanced stability under low-pH operating conditions
Brand:  Grace™ 5110656
Encompass
 View more versions of this product
Product Code. 12778685
  • 738.00 € / Each
Estimated Shipment: 11-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Excellent peak symmetry
  • Exceptional stability from 1 to 10pH
  • Reduced need for mobile-phase modifiers
  • Long column life
  • C18 bonding is monomeric
Specifications
Specifications
3.0 mm
50 mm
C18
Analysis of a Wide Range of Analytes and Impressive Peak Symmetry for Both Acidic and Basic Compounds
4 μm
Column