missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sartorius Stedim Biotech™ Grade 1291 Qualitative Papers, Folded
Sartorius Stedim Biotech™ Grade 1291 Qualitative Papers, Folded are suitable for analytical purposes and routine analyses whenever no gravimetric analyses are required.
Brand: Sartorius Stedim Biotech™ FT-4-209-600
This item is not returnable.
View return policy
Description
- Wet-strengthened.
- Constructed of refined pulp and linters with > 95% alpha-cellulose content.
- Ash content of < 0.1% according to DIN 54370
Specifications
84 g/m2 | |
Cotton Linters, Refined Pulp | |
180 sec. | |
100/Pk. | |
600 mm |
Folded | |
2 to 3 μm | |
1291 | |
0.15 μm |