missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Grade 25 Ruby Spheres, Dia.: 1.19mm
50.00 € - 485.00 €
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11355979
|
Thermo Scientific™
042041.KM |
25 Pieces |
184.00 €
Pack of 25 |
Estimated Shipment: 14-01-2025
Log in to see stock. |
|||||
15446425
|
Thermo Scientific™
042041.KJ |
5 Pieces | N/A | ||||||
11365979
|
Thermo Scientific™
042041.KN |
100 Pieces | N/A | ||||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Ruby Sphere | |
1.19 mm | |
25 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title