missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Grade 605 Very Absorbent Joseph Paper

Used in laboratories and hospitals. Fisherbrand™ Grade 605 Very Absorbent Joseph Paper is thin and not fluffy.

Brand:  Fisherbrand™ 0605F00021

4.95 EUR valid until 2025-05-02
Use promo code "24151" to get your promotional price.


 View more versions of this product

Product Code. 11728722

  • 6.40 € / Pack of 200
In Stock
to see stock.
Edge
Explore more special offers
Add to basket

Description

Description

Applications:
  • Cleaning and drying of, glass containers, tubes, flasks, microscopic plates, bottles
Specifications

Specifications

150 mm
150 x 150 mm
Cleaning and drying of, Glass containers, tubes, flasks, microscopic plates, bottles in laboratories and hospitals
605
150 mm
Sheet
200 Pk.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Fisherbrand™ Grade 605 Very Absorbent Joseph Paper > Sheet; Dimensions:150 x 150mm; Quantity: 200PK

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.