Learn More
Fisherbrand™ Grade 7 Cellulose Fiber and Cotton Linter Extraction Thimble
Suitable for all extractors systems.
Brand:  Fisherbrand™ 0007N00124
 View more versions of this product
Product Code. 11754536
  • 125.00 € / Pack of 25
Estimated Shipment: 31-12-2024
to see stock.
Explore more special offers
Add to basket
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Description
Description
  • 100% High pure cotton linters fibers
  • Neutral and fat free
  • High mechanical strength
  • Excellent retention capacity
Applications:
  • Extraction of sulphur
  • Extraction of the content of free lipids in food
  • Determination of dust and tar in industrial air and gases
  • Lacquer, binder analysis in paint colours
  • Control of emission smokes in industrial chimneys
  • Gravimetric analysis of dust in gases
Specifications
Specifications
43 mm
Extraction of Sulfur in gaseous masses, determination of dust and tar in industrial air and gases
Suitable for all extractors systems
Extraction Thimble
High Purity Cellulose Fibers and Cotton Linters
7
123 mm