missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BUCHI Graduated Glass Sample Tube Set with 0.3mL Residual Volume
Use these tubes for a variety of parallel concentration applications. BUCHI Graduated Glass Sample Tube Set with 0.3mL Residual Volume are designed for use with the Syncore™ R-12 Analyst.
Brand: BUCHI 046072
This item is not returnable.
View return policy
Description
- Set of 12 glass tubes
- For use with Syncore R-12 Analyst
- For working volumes up to 120mL
- Residual volume 0.3mL
Specifications
For Crystal Analyst rack R-12 12 x 120mL | |
Glass |