missing translation for 'onlineSavingsMsg'
Learn More

Honeywell™ Grain Leather Drivers Gloves

Product Code. 12681063
Click to view available options
Size:
8
9
11
This item is not returnable. View return policy
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
This item is not returnable. View return policy
You may only order up to the available stock. Where an alternative item has been identified it will be shown below

Calf grain leather provides comfort and flexibility. The winged thumb and reinforced index finger make the glove more anatomical for additional ease of use.

  • Cowhide grain leather
  • Elasticated back
  • Gunn cut
  • Wing thumb
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance CE CATEGORY II Intermediary risks, 420-03 EEC Basic standards, EN 388-03 Mechanical hazards, EC Category PPE 2, Quality Assurance ISO 9001 / 2000, EC Certificate Number N°0075/797/162/12/06/0358, EC Attestation Number N°0075/797/162/12/06/0358
Certifications/Compliance (Europe) CE Category: II Intermediary Risks, In accordance with : EN 420-03 EEC Basic Standards, and EN 388-16 Mechanical Hazards
Classification Dexterity: 5
Cut Style Gunn
Coating Material Leather
Color White
Cut Resistance Rating 1
Hand Specific
Material Grain Cowhide Leather
Product Type Driver Gloves
Size 11
Style Gunn
For Use With (Application) General Handling in Dry Environments
Glove Cuff Style Open
Includes Elasticized wrist
Lining Light & medium handling
Palm Type Leather
Quantity 10Pair/PK
Series 2030195
Tear Rating 2
Thumb Style Wing
Length (Metric) 25 cm
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'