Learn More
Pro-Lab Diagnostics™ Gram's Iodine Solution
Use for convenient Gram staining of bacteria cells. Prolab Diagnostics™ Gram's Iodine Solution concentrates are 10 times concentrated to give a ready to use stain on dilution.
Brand: Pro-Lab Diagnostics™ PL.7005
This item is not returnable.
View return policy
Description
- Pro-Lab Diagnostics offers an extensive range of ready-to-use and concentrated microbiological staining solutions
- Solutions come in a selection of convenient and economical sizes.
- All stains are manufactured in accordance with all current regulatory requirements
- CE labelled
Specifications
Reagent | |
2L | |
Liquid | |
Dark brown | |
Gram Stain |
Gram Iodine | |
(EC) No 1907/2006, Annex II, (EU) No 453/2010, CE-mark | |
For Gram staining | |
Ethanol |
Product Content Correction
Product Title
Pro-Lab Diagnostics™ Gram's Iodine Solution > Volume: 2L
Spot an opportunity for improvement?Share a Content Correction