missing translation for 'onlineSavingsMsg'
Learn More
Graphene, nanoplatelets 2-10 nm, Thermo Scientific Chemicals Product Code.: 15575886

Graphene, nanoplatelets 2-10 nm, Thermo Scientific Chemicals

Product Code. 15575886
50 g
Click to view available options
Quantity:
50 g
This item is not returnable. View return policy

Product Code. 15575886

Brand: Thermo Scientific Acros 453160500

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

 

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 1034343-98-0
Melting Point 3650.0°C
Molecular Formula Cx
MDL Number MFCD00144065
Quantity 50 g
Synonym methyl hydride, marsh gas, biogas, fire damp, r 50 refrigerant, methan, metano, hsdb 167, in gaseus state, tetrahydridocarbon
Solubility Information Solubility in water: insoluble.
InChI Key VNWKTOKETHGBQD-UHFFFAOYSA-N
SMILES C
IUPAC Name methane
Molecular Weight (g/mol) 16.04
PubChem CID 297
ChEBI CHEBI:16183
Formula Weight 12.01
Chemical Name or Material Graphene
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0467419 Certificate TypeCertificate of Analysis Date25/01/2025 Catalog Number453160500
Lot NumberA0468901 Certificate TypeCertificate of Analysis Date25/01/2025 Catalog Number453160500
Lot NumberA0466413 Certificate TypeCertificate of Analysis Date25/09/2024 Catalog Number453160500
Lot NumberA0463254 Certificate TypeCertificate of Analysis Date11/06/2024 Catalog Number453160500
Lot NumberA0461855 Certificate TypeCertificate of Analysis Date07/05/2024 Catalog Number453160500
Product Identifier
  • Graphene, single layer

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Graphene, nanoplatelets 2-10 nm, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.