missing translation for 'onlineSavingsMsg'
Learn More
Graphite conductive adhesive, aqueous based, Thermo Scientific Chemicals Product Code.: 15411817

Graphite conductive adhesive, aqueous based, Thermo Scientific Chemicals

Product Code. 15411817
30 g
Click to view available options
Quantity:
30 g
This item is not returnable. View return policy

Product Code. 15411817

Brand: Thermo Scientific Alfa Aesar 042466.15

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

 

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Color Black
Density 1.16g/cm3 at 20°C
Ignition Point 455°C (851°F)
Odor Sweet
MDL Number MFCD01866817
Quantity 30 g
Vapor Pressure 23 hPa at 20°C
Chemical Name or Material Graphite conductive adhesive, Aqueous based

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberW10G012 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number042466.15
Lot NumberW29C068 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number042466.15
Lot NumberQ28H022 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog Number042466.15
Lot NumberC03T052 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog Number042466.15
Lot NumberY05E048 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog Number042466.15

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Graphite conductive adhesive, aqueous based, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.