Learn More
Thermo Scientific Chemicals Graphite, Fusion Crucible, drillpoint, unpurified, volume 7.88cc
Manufactured from graphite materials that provide exceptional thermal uniformity and exactness to dimensional specifications.
414.06 € - 462.30 €
Specifications
Description Unpurified extruded graphite
Material Graphite
For Use With (Application) Metallurgy
Product Type Specs Graphite fusion crucible
Shape Drillpoint
View More Specs
Products 2
Product Code Brand Capacity (Metric) Shape Quantity Price Quantity & Availability  
Product Code Brand Capacity (Metric) Shape Quantity Price Quantity & Availability  
15466345
View Documents
Thermo Scientific Chemicals
040794.KD
7.5 cc Drillpoint 50pc
462.30 €
Pack of 50
In Stock
to see stock.
Add to basket
15426055
View Documents
Thermo Scientific Chemicals
014747.UB
7.88 cc Drillpoint 36pc
414.06 €
Pack of 36
In Stock
to see stock.
Add to basket
Description
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Unpurified extruded graphite
Metallurgy
Drillpoint
Graphite
Graphite fusion crucible