Learn More
Graphite plate, 1.27cm (0.5in) thick, Thermo Scientific Chemicals
106.53 € - 315.57 €
Specifications
Chemical Name or Material | Graphite plate |
---|---|
Name Note | 1.27cm (0.5 in.) thick |
MDL Number | MFCD00144065 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11351188
|
Thermo Scientific Alfa Aesar
010132.RI |
12.5 x 25 cm |
106.53 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11361188
|
Thermo Scientific Alfa Aesar
010132.RJ |
25 x 50 cm |
315.57 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Graphite plate | |
1.27cm (0.5 in.) thick | |
MFCD00144065 |
Safety and Handling
missing translation for 'einecsNumber' : 231-955-3
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.