Learn More
Graphite plate, highly oriented pyrolytic graphite (HOPG), 10x10x(1.6min)mm (0.394x0.394x0.079in), Thermo Scientific Chemicals
593.96 € - 2197.94 €
Specifications
Chemical Name or Material | Graphite plate |
---|---|
Name Note | Highly oriented pyrolytic graphite (HOPG), 10x10x(1.6min)mm (0.394x0.394x0.079 in.) |
MDL Number | MFCD00144065 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15423905
|
Thermo Scientific Alfa Aesar
043835.KF |
1 Pc. |
593.96 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15433905
|
Thermo Scientific Alfa Aesar
043835.KJ |
5 Pc. |
2197.94 €
Pack of 5 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Graphite plate | |
Highly oriented pyrolytic graphite (HOPG), 10x10x(1.6min)mm (0.394x0.394x0.079 in.) | |
MFCD00144065 |
Safety and Handling
missing translation for 'einecsNumber' : 231-955-3
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.