Learn More
Graphite plate, pyrolytic, 1.27x9.98x9.98mm (0.05x0.393x.393in), Thermo Scientific Chemicals
Solid Plate for SEM calibration
Brand:  Thermo Scientific Alfa Aesar 040156.KF
 View more versions of this product
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 15466315
  • 255.27 € / Each
Explore more special offers
Description
Description
Graphite plate is used for fuel cells, electrolysis, electroplating, and other various mechanical applications where friction, heat, or electrical properties are factors. It is also used as a solid plate for SEM calibration.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Graphite plate, pyrolytic
MFCD00144065
Insoluble in water.
1.27x9.98x9.98mm (0.05x0.393x.393 in.)
1 Pc.