Learn More
Graphite, powder, Thermo Scientific Chemicals
60.70 € - 60.80 €
Chemical Identifiers
CAS | 7782-42-5 |
---|---|
Molecular Formula | C |
Molecular Weight (g/mol) | 12.01 |
MDL Number | MFCD00133992 MFCD00144065 MFCD00146977 MFCD00211867 MFCD06411993 MFCD07370731 |
InChI Key | OKTJSMMVPCPJKN-UHFFFAOYSA-N |
Synonym | graphite, activated charcoal, norit, mineral, carbon-12, carbono, graphene, acticarbone, anthrasorb, carbosieve |
PubChem CID | 5462310 |
ChEBI | CHEBI:27594 |
IUPAC Name | carbon |
SMILES | [C] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10731335
|
Thermo Scientific Acros
385031000 |
100 g | Glass bottle |
60.70 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10380394
|
Thermo Scientific Acros
385030010 |
1 kg | Glass bottle |
60.80 €
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
AdsorbentChemical Identifiers
7782-42-5 | |
12.01 | |
OKTJSMMVPCPJKN-UHFFFAOYSA-N | |
5462310 | |
carbon |
C | |
MFCD00133992 MFCD00144065 MFCD00146977 MFCD00211867 MFCD06411993 MFCD07370731 | |
graphite, activated charcoal, norit, mineral, carbon-12, carbono, graphene, acticarbone, anthrasorb, carbosieve | |
CHEBI:27594 | |
[C] |
Specifications
7782-42-5 | |
C | |
15, 4575 | |
OKTJSMMVPCPJKN-UHFFFAOYSA-N | |
carbon | |
5462310 | |
12.01 | |
Graphite |
Black | |
MFCD00133992 MFCD00144065 MFCD00146977 MFCD00211867 MFCD06411993 MFCD07370731 | |
graphite, activated charcoal, norit, mineral, carbon-12, carbono, graphene, acticarbone, anthrasorb, carbosieve | |
[C] | |
12.01 | |
CHEBI:27594 | |
Powder |
Safety and Handling
GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Warning
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.