Learn More
Graphite rod, 3.05mm (0.12in) dia x 305mm (12in) long, 99.9995% (metals basis), Thermo Scientific Chemicals
237.00 € - 897.00 €
Specifications
Chemical Name or Material | Graphite rod |
---|---|
Name Note | 3.05mm (0.12 in.) dia. x 305mm (12 in.) long |
Purity Grade Notes | AGKSP grade, ultra 'F' purity |
Assay Percent Notes | (metals basis) |
MDL Number | MFCD00144065 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11430341
|
Thermo Scientific Alfa Aesar
040765.KM |
25 Pc. |
237.00 €
Pack of 25 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11440341
|
Thermo Scientific Alfa Aesar
040765.KN |
100 Pc. |
897.00 €
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Graphite rod is used in computer brushes, anodes, arc lamps, carbon coating, cathode ray tubes, and moderator in nuclear piles. It is used for high-temperature crucibles, as a lubricant and in lead pencils. Graphite rod coated with Th(IV), plays an useful application for ion selective electrode based on thorium oxinate complex. It is also essential for the preparation of lead(II)-selective PVC-coated graphite rod electrode.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Commutar brushes, anodes, arc lamps, carbon coating, cathode ray tubes, moderator in nuclear piles
Specifications
Graphite rod | |
AGKSP grade, ultra 'F' purity | |
MFCD00144065 | |
99.9995% |
3.05mm (0.12 in.) dia. x 305mm (12 in.) long | |
(metals basis) | |
Insoluble in water. Soluble in molten nickel. |
Safety and Handling
missing translation for 'einecsNumber' : 231-955-3
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only