missing translation for 'onlineSavingsMsg'
Learn More

Trajan Scientific and Medical™ Graphite Sealing Ring for PerkinElmer Liner

Made up of graphite and used as sealing rings for PerkinElmer GC.

Brand:  Trajan Scientific and Medical™ 0726522

Product Code. 15208883

  • 58.40 € / Pack of 10
Estimated Shipment: 23-05-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Suitable for PerkinElmer GC
  • Temperature stable up to 450°C
Specifications

Specifications

For use with 4 mm OD liners
Graphite
10/Pk.
Graphite Sealing Ring for PerkinElmer
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Trajan Scientific and Medical™ Graphite Sealing Ring for PerkinElmer Liner > Graphite Sealing Ring for PerkinElmer Liner

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.