missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Green Caps For Lysing Matrix Tubes

Colored screw cap with o-ring fits empty 2 mL microfuge tubes. Green Caps for Lysing Matrix Tubes are used in conjunction for isolation of total RNA from plants and animals.

Brand:  MP Biomedicals™ 115067005

351.49 EUR valid until 2025-01-31
Use promo code "23894" to get your promotional price.


Product Code. 11412350

  • 377.00 € / Pack of 1000
Estimated Shipment: 19-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Fit empty 2 mL FastPrep tubes
  • For use in FastPrep systems
  • Available separately for those who wish to fill there own lysing matrix tubes.
  • Applications include sample preparation, sample lysis, nucleic acid isolation and purification, protein isolation, molecular biology, gene expression, microbiological analysis, gene function, gene regulation
TRUSTED_SUSTAINABILITY
Specifications

Specifications

O-ring
Green
2 mL
Screw Cap
1000 Pk.
FastRNA™ Pro Green Lysing Matrix D Tubes
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.