missing translation for 'onlineSavingsMsg'
Learn More

Prestige Medical Green Gasket

Green Gasket for 121°C & 126°C models.

52.55 € - 80.00 €

Specifications

Product Type Green Gasket
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17879578
View Documents
Prestige Medical
219776
2 Pk. N/A N/A N/A
15805025
View Documents
Prestige Medical
219500
1/Pk.
52.55 €
Each
In Stock
to see stock.
Edge
Add to basket
 
Description

Description

  • Green Gasket for 121°C & 126°C models.
Specifications

Specifications

Green Gasket
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Prestige Medical Green Gasket

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.