missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Deutsch Neumann™ Grey Natural Rubber Spacer
7.60 € - 8.00 €
Product Code | Brand | Height (Metric) | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Height (Metric) | Quantity | Price | Quantity & Availability | ||||
10129090
|
Deutsch Neumann™
2010021 |
20 mm | 10/Pk. |
7.60 €
Pack of 10 |
Estimated Shipment: 07-01-2025
Log in to see stock. |
||||
10747361
|
Deutsch Neumann™
2010031 |
24 mm | 10/Pk. |
8.00 €
Pack of 10 |
Estimated Shipment: 06-01-2025
Log in to see stock. |
||||
Specifications
Gray | |
Natural Rubber |