Learn More
SPEX™ SamplePrep Grinding Vial Accessory Packages
Contains everything necessary to run Small/Mid-size/Large grinding vials in the LargeFreezer/Mill™
Brand:  SPEX™ SamplePrep 6870S
 View more versions of this product
Product Code. 15330502
  • 1155.00 € / 1 set
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Closed grinding vial prevents cross-contamination.
  • Excellent for grinding tissue, seeds, polymers, pharmaceuticals, and textiles.
  • Compatible with grinding vial sets 04576-35 and 61043-07.
  • Compatible with Freezer/Mill™ cryogenic grinders 04500-39 to -42, 04577-77, and 04577-92 to -94.
Specifications
Specifications
Grinding Set
SamplePrep™ Grinding Vial, Vial Set
1 Multivial Holder (04500-51), 1 Vial Extractor/Opener (04576-31), and 1 Vial Rack (04576-32)
Small