Learn More
Ground & Waste Water Pollution Standard Solution 4, Specpure™, Thermo Scientific Chemicals
240.20 € - 573.00 €
Specifications
Chemical Name or Material | Ground & Waste Water Pollution Standard Solution 4 |
---|---|
MDL Number | MFCD03792306 |
UN Number | UN3264 |
Grade | Specpure™ |
Physical Form | Liquid |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11329198
|
Thermo Scientific Alfa Aesar
042579.AE |
100 mL |
240.20 €
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11336119
|
Thermo Scientific Alfa Aesar
042579.AP |
500 mL | N/A | ||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Ground & Waste Water Pollution Standard Solution 4 | |
UN3264 | |
Liquid |
MFCD03792306 | |
Specpure™ |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P234-P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362-P390
H290-H315-H319
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, INORGANIC, N.O.S.
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.