missing translation for 'onlineSavingsMsg'
Learn More

Ground & Waste Water Pollution Standard Solution 5, Specpure™, Thermo Scientific Chemicals

150.75 € - 356.00 €

Specifications

Chemical Name or Material Ground & Waste Water Pollution Standard Solution 5
MDL Number MFCD03792307
UN Number UN3264
Grade Specpure™
Physical Form Liquid
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11339198
View Documents
Thermo Scientific Alfa Aesar
042580.AE
100 mL
150.75 €
100mL
Please to purchase this item. Need a web account? Register with us today!
11346119
View Documents
Thermo Scientific Alfa Aesar
042580.AP
500 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Ground & Waste Water Pollution Standard Solution 5
UN3264
Liquid
MFCD03792307
Specpure™
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P201-P202-P210-P222-P223-P231+P232-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P281-P302+P334-P304+P340-P305+P351+P338-P310-P330-P331-P335+P334-P363-P370+P378q-P501c

H225-H250-H260-H304-H314-H335-H336-H361f-H373

missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, INORGANIC, N.O.S.

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only