missing translation for 'onlineSavingsMsg'
Learn More

GSK 4716, Thermo Scientific™

Product Code. 15436769
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Selective agonist at estrogen-related receptors ERR→ and ERR→

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 101574-65-6
Molecular Formula C17H18N2O2
Molecular Weight (g/mol) 282.34
MDL Number MFCD00567155
InChI Key IKPPIUNQWSRCOZ-WOJGMQOQSA-N
Synonym e-4-hydroxy-n'-4-isopropylbenzylidene benzohydrazide, 4-hydroxy-benzoic acid 4-isopropyl-benzylidene-hydrazide, 4-hydroxy-2-1e-4-1-methylethyl phenyl methylene hydrazide, 2-4-1-methylethyl phenyl methylene hydrazide 4-hydroxybenzoic acid, 2gpp, d0dx8t, gsk hplc, 4-hydroxy-n-e-4-isopropylphenyl methyleneamino benzamide, 4-hydroxy-n-e-4-propan-2-ylphenyl methylideneamino benzamide, 4-hydroxy-n-1e-4-propan-2-yl phenyl methylidene benzohydrazide
PubChem CID 5331325
ChEBI CHEBI:80000
IUPAC Name 4-hydroxy-N-[(E)-(4-propan-2-ylphenyl)methylideneamino]benzamide
SMILES CC(C)C1=CC=C(\C=N\NC(=O)C2=CC=C(O)C=C2)C=C1

Specifications

Color White
Quantity 10 mg
UN Number UN3077
Formula Weight 282.34
Physical Form Solid
Chemical Name or Material GSK 4716
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'