Learn More
Tocris Bioscience™ GSK 650394
Serum- and glucocorticoid-regulated kinase (SGK) inhibitor
Brand:  Tocris Bioscience™ 3572/50
972.98 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.
 View more versions of this product
Product Code. 18044002
  • 1240.00 € / 50mg
Explore more special offers
Description
Description
GSK 650394 is a serum- and glucocorticoid-regulated kinase 1 (SGK1) inhibitor (IC50 values are 62 and 103 nM for SGK1 and SGK2 respectively). Displays >30-fold selectivity over Akt and other related kinases. Inhibits androgen-stimulated growth of LNCaP cells, a human prostate carcinoma cell line (IC50 ∽ 1 μM).
Specifications
Specifications
>97%
GSK 650394
GSK 650394
50 mg
382.45