Learn More
Tocris Bioscience™ GSK 650394
Serum- and glucocorticoid-regulated kinase (SGK) inhibitor
Brand:  Tocris Bioscience™ 3572/10
231.53 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.
 View more versions of this product
Product Code. 18789796
  • 303.00 € / 10mg
Explore more special offers
Description
Description
GSK 650394 is a serum- and glucocorticoid-regulated kinase 1 (SGK1) inhibitor (IC50 values are 62 and 103 nM for SGK1 and SGK2 respectively). Displays >30-fold selectivity over Akt and other related kinases. Inhibits androgen-stimulated growth of LNCaP cells, a human prostate carcinoma cell line (IC50 ∽ 1 μM).
Specifications
Specifications
>97%
GSK 650394
GSK 650394
10 mg
382.45