missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GSK J4, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
Histone lysine demethylase (KDM) inhibitor; blocks demethylation of histone H3K27. Attenuates lipopolysaccharide (LPS)-induced proinflammatory cytokine production in primary human macrophages (IC50 = 9 μM for the inhibition of TNFα release). Cell permeable. Ethyl ester derivative of GSK J1 .
Negative Control also available.
Specifications
Specifications
Quantity | 10 mg |
Formula Weight | 417.5 |
Percent Purity | >99% |
Chemical Name or Material | GSK J4 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction