missing translation for 'onlineSavingsMsg'
Learn More

GSK369796 Dihydrochloride, MedChemExpress

MedChemExpress GSK369796 Dihydrochloride is an affordable and effective antimalarial and inhibits hERG potassium ion channel repolarization with an IC50 of 7.5 μM.

Encompass_Preferred

183.60 € - 1841.40 €

Specifications

Chemical Name or Material GSK369796 Dihydrochloride
Physical Form Solid
CAS 1010411-21-8
Color Yellow
Purity Grade Notes Research
View More Specs

Products 5
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16490146
Encompass_Preferred
View Documents
Medchem Express
HY-12082A
2 mg
183.60 €
2mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16484978
Encompass_Preferred
View Documents
Medchem Express
HY-12082A
10 mg
368.55 €
10mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16475746
Encompass_Preferred
View Documents
Medchem Express
HY-12082A
100 mg
1841.40 €
100mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16451227
Encompass_Preferred
View Documents
Medchem Express
HY-12082A
50 mg
1104.30 €
50mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16407717
Encompass_Preferred
View Documents
Medchem Express
HY-12082A
5 mg
267.30 €
5mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

GSK369796 Dihydrochloride is an affordable and effective antimalarial and inhibits hERG potassium ion channel repolarization with an IC50 of 7.5 μM.
Specifications

Specifications

GSK369796 Dihydrochloride
1010411-21-8
Research
COVID-19-immunoregulation
OC1=CC(NC2=CC=NC3=CC(Cl)=CC=C23)=CC=C1CNC(C)(C)C.[H]Cl.[H]Cl
428.78
Research
Solid
Yellow
C20H24Cl3N3O
DMSO : 100 mg/mL (233.22 mM; Need ultrasonic) ∣H2O : 50 mg/mL (116.61 mM; Need ultrasonic)
428.78
98.32%
Videos
Safety and Handling

Safety and Handling

H315∣H319∣H335

missing translation for 'shelfLife' : 4°C, sealed storage, away from moisture∣In solvent : -80°C, 6 months∣-20°C, 1 month (sealed storage, away from moisture)

Recommended Storage : 4°C, sealed storage, away from moisture∣In solvent : -80°C, 6 months∣-20°C, 1 month (sealed storage, away from moisture)

SDS
Documents

Documents

Product Certifications

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title
GSK369796 Dihydrochloride, MedChemExpress

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.