Learn More
Cytiva GST Bulk Kit
For small-scale manual purification of GST-tagged proteins using either column chromatography or batch method. Facilitates optimization studies of expression, solubility conditions and purification parameters of GST-tagged proteins
Brand:  Cytiva 27-4570-01
Product Code. 10166044
  • 946.00 € / Each
Estimated Shipment: 14-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Gentle elution conditions preserve protein antigenicity and function
  • Flexible GST-glutathione affinity system for rapid column or batch purification
  • High specificity (compared to histidine-tagged purification)
  • Provide reagents for induction expression and elution of fusion proteins
  • Sufficient reagents to purify up to 50mg fusion protein
  • Includes 10mL bulk pack of Glutathione Sepharose™ 4B, 5 disposible columns, GST buffer kit and 500 mg Isopropyl Β-D Thiogalactoside
TRUSTED_SUSTAINABILITY
Specifications
Specifications
GST bulk kit is designed for small-scale manual purification of GST-tagged proteins using either column chromatography or batch method. Facilitates optimization studies of expression, solubility conditions and purification parameters of GST-tagged protein
20°C to 25°C
Chromatography Media
10 mL bulk pack of Glutathione Sepharose 4B medium, 5 x gravity-flow columns, IPTG, GST Buffer Kit
1 Kit