missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva GSTrap™ FF Columns: 16 x 25mm
Purifies GST-tagged proteins in one step
Brand: Cytiva 17-5131-02
1330.95 EUR valid until 2025-02-21
Use promo code "24115" to get your promotional price.
This item is not returnable.
View return policy
Description
- Mild elution conditions preserve antigenicity and function of proteins extracted
- Prepacked with Glutathione Sepharose™ media
- Average particle size: 90μm
- Bed dimensions: 16 × 25mm
- Flow velocity: <20mL/min.
- Matrix: Highly cross-linked 4% agarose
- Ligand: glutathione and 10-carbon linker arm
- Binding capacity: >10mg/mL
- pH stability working range: 3 to 12
Specifications
5 mL | |
16 mm | |
<20 mL/min. | |
3 to 12 | |
4°C to 30°C, 20% Ethanol | |
90 μm | |
For the purification of GST-tagged proteins from bacterial lysates and other glutathione S-tranferases or glutathione-dependent proteins and for preparation of cytoplasmic extract | |
Glutathione and 10 carbon linker arm |
0.62 in. | |
GSTrap FF columns are prepacked Glutathione Sepharose Fast Flow columns for fast, convenient, one-step purification of glutathione S-transferase (GST) tagged proteins. | |
Polypropylene | |
4% Highly Cross-linked Agarose | |
5 bar (0.5 MPa, 70 psi) | |
5 x 5 mL | |
Pre-packed Column |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- GSTrap 5 ml, 5 x 5 ml
Signal Word
- Warning
Hazard Category
- Flammable liquid Category 3
Hazard Statement
- H226-Flammable liquid and vapour.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P235-Keep cool.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : ethanol
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction