missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva GSTrap™ HP Columns
Use to provide high-resolution, one-step purifications of glutathione S-Transferase (GST) tagged proteins. Cytiva Life Sciences™ GSTrap™ HP Columns are prepacked with Glutathione Sepharose™ High Performance medium.
390.00 € - 1587.00 €
Specifications
Description | GSTrap HP columns are prepacked with Glutathione Sepharose High Performance resin and provide high-resolution, one-step purifications of glutathione S-Transferase (GST) tagged proteins. |
---|---|
Material | Polypropylene |
pH Range | 3 to 12 |
Support Type | 4% Highly Cross-linked Agarose |
Content And Storage | 4°C to 30°C, 20% Ethanol |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10647455
|
Cytiva
17-5281-01 |
5 x 1 mL |
413.00 €
Pack of 5 |
In Stock
Log in to see stock. |
|||||
11565035
|
Cytiva
17-5282-02 |
5 x 5 mL |
1587.00 €
Pack of 5 |
In Stock
Log in to see stock. |
|||||
11713219
|
Cytiva
17-5282-01 |
1 x 5 mL |
390.00 €
Each |
In Stock
Log in to see stock. |
|||||
Description
- One-step purification of GST-tagged proteins and other glutathione S-transferases or glutathione-dependent proteins
- Glutathione coupled to Sepharose™ High Performance media
- Column material: polypropylene
- May be used with syringe, peristaltic pump or chromatography system
- Purification process preserves antigenicity and function
- Available as 1mL and 5mL columns
- Average particle size: 34μm
- Ligand: Glutathione
- Matrix: Highly cross-linked 6% agarose
- Binding capacity: >7mg/mL
- pH stability working range: 3 to 12
1mL columns
- Bed dimensions: 7 x 25mm
- Flow velocity: <4mL/min.
- Bed dimensions: 16 x 25mm
- Flow velocity: <20mL/min.
Specifications
GSTrap HP columns are prepacked with Glutathione Sepharose High Performance resin and provide high-resolution, one-step purifications of glutathione S-Transferase (GST) tagged proteins. | |
3 to 12 | |
4°C to 30°C, 20% Ethanol | |
34 μm | |
Pre-packed Column |
Polypropylene | |
4% Highly Cross-linked Agarose | |
5 bar (0.5 MPa, 70 psi) | |
Glutathione S-transferase (GST) tagged proteins produced using the pGEX series of expression vectors, Other glutathione S-transferases and glutathione-dependent proteins | |
Glutathione |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- GSTrap HP 1 ml, 5 x 1 ml
Signal Word
- Warning
Hazard Category
- Flammable liquid Category 3
Hazard Statement
- H226-Flammable liquid and vapour.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P235-Keep cool.
- P501-Dispose of contents/container to .
Supplemental information
- MIXTURE LIST-Contains : ethanol
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title