Learn More
Fisherbrand™ GT2 Benchtop Centrifuge Packs
The mid-sized Fisherbrand™ GT2 Benchtop Centrifuge provides speed and versatility, reaching 4238xg for fast and efficient separations
Brand:  Fisherbrand™ 50156857
Product Code. 15818722
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Mid-size benchtop centrifuge with speed (reaching 4238xg) and versatility but with minimal lab footprint
  • Ventilated and refrigerated versions available, with rapid temperature pull-downs
  • Includes a TX-400 rotor with adapters accommodating 36x15mL or 16x50mL conical tubes, ideal for cell culture applications
  • One-touch, easy-to-use operation with pre-saved protocols
  • Backlit display interface with multi-language options
  • Rapid push-button rotor exchange with the Auto-Lock™ rotor swap system
  • Corrosion resistant and lubricating rotor-cross coating
  • Fitted with ClickSeal™ biocontainment lids
  • Glove and wipe-clean, detergent-friendly
  • Two year warranty
TRUSTED_SUSTAINABILITY
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products