Learn More
Guanidine carbonate, 98%, Thermo Scientific Chemicals
Beschrijving
It is employed as a strong organic alkali, organic intermediate, in soap and cosmetic products, and in the textile industry. It is also used to make fine chemicals and in consumer hair relaxer products. It is used in various industries to solve different purpose such as cosmetics as a hair relaxer that straightens hair by chemically uncurling it, detergents as a synergistic agent, pharmaceuticals as an antibiotic and microencapsulation process for carbonless paper.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
It is employed as a strong organic alkali, organic intermediate, in soap and cosmetic products, and in the textile industry. It is also used to make fine chemicals and in consumer hair relaxer products. It is used in various industries to solve different purpose such as cosmetics as a hair relaxer that straightens hair by chemically uncurling it, detergents as a synergistic agent, pharmaceuticals as an antibiotic and microencapsulation process for carbonless paper.
Solubility
Soluble in water 450 g/L (20°C); slightly soluble in alcohol and acetone.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specificatie
Specificatie
Melting Point | ∼198°C (decomposition) |
Quantity | 250 g |
Beilstein | 3628359 |
Solubility Information | Soluble in water 450g/L (20°C); slightly soluble in alcohol and acetone. |
Formula Weight | 180.17 |
Percent Purity | 98% |
Chemical Name or Material | Guanidine carbonate |
RUO – Research Use Only