missing translation for 'onlineSavingsMsg'
Learn More
Guanidine hydrochloride, 98%, Thermo Scientific Chemicals Product Code.: 11420024

Guanidine hydrochloride, 98%, Thermo Scientific Chemicals

Product Code. 11420024
250 g
Click to view available options
Quantity:
250 g
1000 g
5000 g
This item is not returnable. View return policy

Product Code. 11420024

Brand: Thermo Scientific Alfa Aesar A13543.30

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Guanidine hydrochloride, CAS # 50-01-1, is an ionic buffer component used for nucleic acids (DNA and RNA) and protein isolation.

Guanidine hydrochloride is used for purification of proteins and nucleic acids. It can be used as a medicine, organic synthesis intermediate and is used in dyes. It acts as an intermediate in the preparation of sulfadiazine, which is an important raw material for sulfamethyldiazine, sulfamethazine and folic acid. It is utilized in the synthesis of 2-amino-pyrimidine, 2- amino -6- methyl-pyrimidine and 2-amino -4,6 - dimethyl-pyrimidine. Also used in RNA isolation to dissociate nucleoproteins and inhibit RNase.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Guanidine hydrochloride is the hydrochloride salt of guanidine, an acetylcholine releasing agent
  • It is a normal product of protein metabolism and is sometimes used for pyrimidine synthesis

Application

  • Guanidine hydrochloride is a powerful denaturant, useful for taking nucleic acids (DNA and RNA) and proteins out of their native conformation
  • Guanidine hydrochloride acts as a strong organic base at physiological pH
  • It is suitable for use in protein and nucleic acid purification protocols
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50-01-1
Molecular Formula CH6ClN3
Molecular Weight (g/mol) 95.53
MDL Number MFCD00013026
InChI Key PJJJBBJSCAKJQF-UHFFFAOYSA-N
Synonym guanidine hydrochloride, guanidinium chloride, guanidine, monohydrochloride, aminoformamidine hydrochloride, aminomethanamidine hydrochloride, guanidine monohydrochloride, guanidine hcl, guanidine chloride, iminourea hydrochloride, carbamidine hydrochloride
PubChem CID 5742
ChEBI CHEBI:32735
IUPAC Name guanidine;hydrochloride
SMILES C(=N)(N)N.Cl

Specifications

Melting Point 178°C to 185°C
Density 1.344
Quantity 250 g
Beilstein 3591990
Sensitivity Hygroscopic
Merck Index 144562
Solubility Information Soluble in water and ethanol.
Formula Weight 95.53
Percent Purity 98%
Chemical Name or Material Guanidine hydrochloride

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number62500196 Certificate TypeCertificate of Analysis Date10/04/2025 Catalog NumberA13543.0B,A13543.30,A13543.0I
Lot Number62500041 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberA13543.0B,A13543.30,A13543.0I
Lot Number5001Q12S Certificate TypeCertificate of Analysis Date27/01/2025 Catalog NumberA13543.30
Lot Number10253482 Certificate TypeCertificate of Analysis Date18/12/2024 Catalog NumberA13543.0B,A13543.30,A13543.0I
Lot Number10251491 Certificate TypeCertificate of Analysis Date30/08/2024 Catalog NumberA13543.0B,A13543.30,A13543.0I
compliance-icons
Product Identifier
  • Guanidine hydrochloride
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Guanidine hydrochloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Guanidine hydrochloride, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.