missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals Guanidine hydrochloride, 99+% Product Code.: 15443499

Thermo Scientific Chemicals Guanidine hydrochloride, 99+%

Product Code. 15443499
1 kg
Click to view available options
Quantity:
100 g
500 g
1 kg
This item is not returnable. View return policy

Product Code. 15443499

Brand: Thermo Scientific Chemicals J60786.A1

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Guanidine hydrochloride is used for purification of proteins and nucleic acids. It can be used as a medicine, organic synthesis intermediate and is used in dyes. It acts as an intermediate in the preparation of sulfadiazine, which is an important raw material for sulfamethyldiazine, sulfamethazine and folic acid. It is utilized to synthesis of 2-amino-pyrimidine, 2- amino -6- methyl-pyrimidine and 2-amino -4,6 - dimethyl-pyrimidine. Also used in RNA isolation to dissociate nucleoproteins and inhibit RNase.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Guanidine hydrochloride is used for purification of proteins and nucleic acids. It can be used as a medicine, organic synthesis intermediate and is used in dyes. It acts as an intermediate in the preparation of sulfadiazine, which is an important raw material for sulfamethyldiazine, sulfamethazine and folic acid. It is utilized to synthesis of 2-amino-pyrimidine, 2- amino -6- methyl-pyrimidine and 2-amino -4,6 - dimethyl-pyrimidine. Also used in RNA isolation to dissociate nucleoproteins and inhibit RNase.

Solubility
Freely soluble in water.Soluble in water and ethanol.

Notes
Hygroscopic. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.May agglomerate upon storage.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50-01-1
Molecular Formula CH6ClN3
Molecular Weight (g/mol) 95.53
MDL Number MFCD00013026
InChI Key PJJJBBJSCAKJQF-UHFFFAOYSA-N
Synonym guanidine hydrochloride, guanidinium chloride, guanidine, monohydrochloride, aminoformamidine hydrochloride, aminomethanamidine hydrochloride, guanidine monohydrochloride, guanidine hcl, guanidine chloride, iminourea hydrochloride, carbamidine hydrochlorideShow MoreShow Less
PubChem CID 5742
ChEBI CHEBI:32735
IUPAC Name guanidine;hydrochloride
SMILES C(=N)(N)N.Cl

Specifications

Melting Point 185°C to 188°C
Color White
Density 1.344 g/mL
Quantity 1 kg
Beilstein 3591990
Sensitivity Hygroscopic
Merck Index 14,4562
Solubility Information Freely soluble in water;Soluble in water and ethanol.
Formula Weight 95.53
Percent Purity ≥99%
Physical Form Powder
Chemical Name or Material Guanidine Hydrochloride
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberQ08L031 Certificate TypeCertificate of Analysis Date10/04/2025 Catalog NumberJ60786.A1,J60786.22,J60786.36
Lot NumberX15K002 Certificate TypeCertificate of Analysis Date25/10/2024 Catalog NumberJ60786.22,J60786.A1,J60786.36
Lot NumberR10K036 Certificate TypeCertificate of Analysis Date17/05/2024 Catalog NumberJ60786.22,J60786.A1,J60786.36
Lot NumberM24K006 Certificate TypeCertificate of Analysis Date31/01/2024 Catalog NumberJ60786.22,J60786.A1,J60786.36
Lot NumberW27J008 Certificate TypeCertificate of Analysis Date06/10/2023 Catalog NumberJ60786.22,J60786.A1,J60786.36
compliance-icons
Product Identifier
  • Guanidine hydrochloride
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Guanidine, hydrochloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals Guanidine hydrochloride, 99+% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.