missing translation for 'onlineSavingsMsg'
Learn More

Guanidine hydrochloride, 99.5+%, Molecular Biology Grade, Thermo Scientific Chemicals

26.86 € - 874.00 €

Chemical Identifiers

CAS 50-01-1
Molecular Formula CH6ClN3
Molecular Weight (g/mol) 95.53
MDL Number MFCD00013026
InChI Key PJJJBBJSCAKJQF-UHFFFAOYSA-N
Synonym guanidine hydrochloride, guanidinium chloride, guanidine, monohydrochloride, aminoformamidine hydrochloride, aminomethanamidine hydrochloride, guanidine monohydrochloride, guanidine hcl, guanidine chloride, iminourea hydrochloride, carbamidine hydrochloride
PubChem CID 5742
ChEBI CHEBI:32735
IUPAC Name guanidine;hydrochloride
SMILES C(=N)(N)N.Cl
View More Specs

Products 4
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
15835228
View Documents Promotion Details
Thermo Scientific
J75823.22
100 g Plastic bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15845228
View Documents Promotion Details
Thermo Scientific
J75823.36
500 g Plastic bottle
196.00 €
500g
Please to purchase this item. Need a web account? Register with us today!
15815368
View Documents Aktionsdetails
Thermo Scientific
J75823.A1
1 kg Plastic bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15825368
missing translation for 'viewDocuments' Aktionsdetails
Thermo Scientific
J75823.A7
5 kg Poly pail
874.00 €
5kg
Please to purchase this item. Need a web account? Register with us today!
Beschreibung

Beschreibung

  • Used for extraction of RNA.
  • Preparation of high molecular weight genomic DNA.
  • Solubilizing inclusion bodies.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Used for extraction of RNA
  • Preparation of high molecular weight genomic DNA
  • Solubilizing inclusion bodies
Spezifikation

Chemische Identifikatoren

50-01-1
95.53
PJJJBBJSCAKJQF-UHFFFAOYSA-N
5742
guanidine;hydrochloride
CH6ClN3
MFCD00013026
guanidine hydrochloride, guanidinium chloride, guanidine, monohydrochloride, aminoformamidine hydrochloride, aminomethanamidine hydrochloride, guanidine monohydrochloride, guanidine hcl, guanidine chloride, iminourea hydrochloride, carbamidine hydrochloride
CHEBI:32735
C(=N)(N)N.Cl

Spezifikation

50-01-1
1.344
(NH2)2C=NH·HCl
3591990
PJJJBBJSCAKJQF-UHFFFAOYSA-N
guanidine;hydrochloride
5742
95.53
Molecular Biology
Guanidine Hydrochloride
185°C to 188°C
CH6ClN3
MFCD00013026
guanidine hydrochloride, guanidinium chloride, guanidine, monohydrochloride, aminoformamidine hydrochloride, aminomethanamidine hydrochloride, guanidine monohydrochloride, guanidine hcl, guanidine chloride, iminourea hydrochloride, carbamidine hydrochloride
C(=N)(N)N.Cl
95.53
CHEBI:32735
≥99.5%
Crystals
Videos
Sicherheit und Handhabung

Sicherheit und Handhabung

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H332-H315-H319-H500

missing translation for 'einecsNumber' : 200-002-3

missing translation for 'rtecsNumber' : MF4300000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate

RUO – Research Use Only