missing translation for 'onlineSavingsMsg'
Learn More

Guanidine Thiocyanate (White Crystalline Powder), Fisher BioReagents™

For the isolation of intact RNA from cell extracts

43.50 € - 114.90 €

Chemical Identifiers

CAS 593-84-0
Molecular Formula C2H6N4S
Synonym Guanidinium Isothiocyanate
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10285253
SDS View Documents
Fisher BioReagents
BP221-250
250 g Glass Bottle
43.50 €
250g
Please to purchase this item. Need a web account? Register with us today!
10503345
SDS View Documents
Fisher BioReagents
BP221-1
1 kg Poly Bottle
114.90 €
1kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

For the isolation of intact RNA from cell extracts

A more powerful denaturing agent than Guanidine Hydrochloride, Guanidine Thiocyanate is suitable for the isolation of intact RNA from cell extracts.
Specifications

Chemical Identifiers

593-84-0
C2H6N4S
Guanidinium Isothiocyanate

Specifications

593-84-0
118°C
5
C2H6N4S
Guanidinium Isothiocyanate
118.16
≥99.0%
Guanidine Thiocyanate
0.8 max. (70% in water at 25°C) at 280nm
White
≥99.0 %
H2NC(:NH)NH2·HSCN
Soluble in water
Negligible
Powder or Solid
Videos
Safety and Handling

Safety and Handling

Emergency Overview
Causes burns by all exposure routes. Harmful by inhalation, in contact with skin and if swallowed. Contact with acids liberates toxic gas. Harmful to aquatic organisms, may cause long-term advers

Signal Word: DANGER!

Recommended Storage : RT

Product Identifier
  • Guanidine thiocyanate

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 C

Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H412-Harmful to aquatic life with long lasting effects.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
  • EUH032-Contact with acids liberates very toxic gas.

Precautionary Statement
  • P233-Keep container tightly closed.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.