missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Guanosine-5'-diphosphate disodium salt
Click to view available options
Quantity:
100 mg
500 mg
Description
Guanosine-5'-diphosphate disodium salt is used to study the kinetics and characteristics of GTPases such as those associated with G-protein coupled receptors. It is also used to study cell signaling processes mediate by guanine nucleotide exchange factors.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Guanosine-5′-diphosphate disodium salt is used to study the kinetics and characteristics of GTPases such as those associated with G-protein coupled receptors. It is also used to study cell signaling processes mediate by guanine nucleotide exchange factors.
Solubility
Soluble in water
Notes
Incompatible with strong oxidizing agents.
Guanosine-5′-diphosphate disodium salt is used to study the kinetics and characteristics of GTPases such as those associated with G-protein coupled receptors. It is also used to study cell signaling processes mediate by guanine nucleotide exchange factors.
Solubility
Soluble in water
Notes
Incompatible with strong oxidizing agents.
Specifications
Color | White |
Quantity | 500 mg |
Solubility Information | Soluble in water |
Formula Weight | 487.17 |
Physical Form | Powder |
Chemical Name or Material | Guanosine-5'-diphosphate disodium salt |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'