Learn More
OHAUS™ Guardian 5000 Hotplate-Stirrer
Reliable hotplate stirrers with exceptional safety features for everyday use.
Brand: OHAUS™ 30500563
565.25 EUR valid until 2024-12-31
Use promo code "21911" to get your promotional price.
This item is not returnable.
View return policy
Description
- An industry-leading early detection system that uses two independent safety controls to continuously monitor the electronics and shut off heating before an overtemperature condition occurs.
- Precise stirring from 60 to 1600 rpm is made possible by the design of a powerful motor, strong magnet and software controlled ramp rate, ensuring secure magnetic coupling for viscous applications.
- The bright LCD display includes large temperature and speed settings, intuitive icons and a prominent hot top indicator - all designed to quickly and easliy monitor performance from across the lab.
Specifications
Aluminum | |
2.812 kg | |
e-G51HSRDM | |
126.000 mm | |
5.00 in. | |
173.00 mm | |
6.80 in. | |
2.600 kg |
230 V | |
230 V, 4.6 A, 50/60 Hz | |
10.50 in. | |
+ 5°C – 380°C | |
50/60 Hz | |
267.00 mm | |
5.700 lb | |
EU |