Learn More
Labconco™ Guardian™ Digital Airflow Monitor Kit
For use with Labconco Basic™ Laboratory Hoods. Labconco™ Guardian™ Digital Airflow Monitor Kit includes audible/visual alarm to alert user to low or high face velocity.
Brand:  Labconco™ 9743209
Product Code. 15680125
  • 3770.00 € / Each
Explore more special offers
Description
Description
  • LCD displays average face velocity
  • Audible/visual alarm alerts user to low or high face velocity
  • For installation in a factory-prepared cutout on upper portion of right side fixture panel
  • For use with Labconco Basic™ Laboratory Hoods
Specifications
Specifications
Airflow Monitor