missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Guinea pig anti-Rabbit IgG whole molecule, Polyclonal, Secondary Antibody, Abnova™
Brand: Abnova PAB9407.1mg
This item is not returnable.
View return policy
Specifications
Guinea pig anti-Rabbit IgG whole molecule | |
Polyclonal | |
Guinea pig anti-Rabbit IgG recognizes Rabbit IgG whole molecule. This secondary antibody was purified using antigen affinity chromatography. | |
In 20mM KH2PO4, 150mM NaCl, pH 7.2 (0.01% sodium azide) | |
Rabbit IgG whole molecule | |
1 mg | |
No reaction was observed against goat, human and mouse serum proteins. | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
ELISA, Immunohistochemistry, Western Blot | |
Unconjugated | |
The optimal working dilution should be determined by the end user. | |
Guinea Pig | |
This product was prepared from monospecific antiserum by immunoaffinity chromatography, followed by solid phase adsorption(s) to remove any unwanted reactivities. | |
Secondary | |
Rabbit | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction