missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cole-Parmer™ Guzzler Diaphragm Hand Pump
120.00 € - 158.00 €
Product Code | Brand | Flow Rate | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Flow Rate | Price | Quantity & Availability | ||||||
11705698
|
Cole-Parmer™
07090-10 |
8.5 gal./min. |
120.00 €
Each |
Estimated Shipment: 29-05-2025
Log in to see stock. |
||||||
11715698
|
Cole-Parmer™
07090-11 |
12 gal./min. |
158.00 €
Each |
Estimated Shipment: 29-05-2025
Log in to see stock. |
||||||
Specifications
101.6 mm | |
254 mm |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Cole-Parmer™ Guzzler Diaphragm Hand Pump