Learn More
GW3965 hydrochloride, MedChemExpress
MedChemExpress GW3965 hydrochloride is a potent and selective liver X receptor (LXR) agonist with EC50s of 190 nM and 30 nM for hLXRα and hLXRβ, respectively.
Brand:  Medchem Express HY-10627A
Encompass_Preferred
 View more versions of this product
Product Code. 16406787
  • 95.85 € / 5mg
Explore more special offers
Description
Description
GW3965 hydrochloride is a potent and selective liver X receptor (LXR) agonist with EC50s of 190 nM and 30 nM for hLXRα and hLXRβ, respectively.
Specifications
Specifications
GW3965 hydrochloride
White
C33H32Cl2F3NO3
DMSO : 100 mg/mL (161.68 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (ultrasonic;warming;heat to 60°C) (insoluble)
618.51
98.13%
Solid
405911-17-3
Research
5 mg
ClC1=C(C(F)(F)F)C=CC=C1CN(CC(C2=CC=CC=C2)C3=CC=CC=C3)CCCOC4=CC(CC(O)=O)=CC=C4.[H]Cl
618.51
Research
COVID-19-immunoregulation