missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ H 151

STING antagonist

251.00 € - 899.00 €

Specifications

Chemical Name or Material N-(4-Ethylphenyl)-N'-1H-indol-3-yl-urea
CAS 941987-60-6
Synonym ERIS, hMITA, MITA, mitochondrial mediator of IRF3 activation, MPYS, NET23, N-terminal methionine-proline-tyrosine-serine plasma membrane tetraspanner, sting, transmembrane protein 173
Target STING Antagonists
Molecular Formula C17H17N3O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18717606
View Documents
Tocris Bioscience™
6675/10
10 mg
251.00 €
10mg
Please to purchase this item. Need a web account? Register with us today!
18769306
View Documents
Tocris Bioscience™
6675/50
50 mg
899.00 €
50mg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • Covalently binds STING at Cys91, blocking palmitoylation and preventing assembly of STING clusters
  • Decreases inflammatory cytokine production in human and mouse cells in vitro
  • Also blocks CMA (Cat. No. 6677) -induced increases in levels of inflammatory cytokines in mice, and alleviates systemic inflammation in a mouse model of autoinflammatory disease
Specifications

Specifications

N-(4-Ethylphenyl)-N'-1H-indol-3-yl-urea
ERIS, hMITA, MITA, mitochondrial mediator of IRF3 activation, MPYS, NET23, N-terminal methionine-proline-tyrosine-serine plasma membrane tetraspanner, sting, transmembrane protein 173
C17H17N3O
98%
941987-60-6
STING Antagonists
279.34
Videos
Safety and Handling

Safety and Handling

Recommended Storage : Store at 4°C

SDS
Documents

Documents

Product Certifications